Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204178.1 | NP_989509.1 | 5 - 6 | 485 | 131 | |
XM_025155402.1 | XP_025011170.1 | 4 - 5 | 1653 | 131 | ||
GenBank mRNA | U18261.1 | - | 5 - 6 | 485 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |