Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCND2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204279.1 | NP_989610.1 | 3 - 4 | 1377 | 69 | |
XM_015276625.2 | XP_015132111.1 | 3 - 4 | 69 | |||
GenBank mRNA | AF075160.1 | - | 3 - 4 | 1377 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |