Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204324.1 | NP_989655.1 | 9 - 10 | 1241 | 111 | |
XM_015295659.2 | XP_015151145.1 | 9 - 10 | 1222 | 111 | ||
XM_015295660.1 | XP_015151146.1 | 10 - 11 | 1242 | 111 | ||
XM_015295661.2 | XP_015151147.1 | 10 - 11 | 1410 | 111 | ||
XM_025141880.1 | XP_024997648.1 | 11 - 12 | 8121 | 111 | ||
XM_025141881.1 | XP_024997649.1 | 11 - 12 | 8146 | 111 | ||
GenBank mRNA | AF113946.1 | - | 9 - 10 | 1241 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |