Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SURF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204355.1 | NP_989686.1 | 2 - 3 | 245 | 88 | |
GenBank mRNA | AB050010.1 | - | 2 - 3 | 245 | 88 | |
BX934974.2 | - | 2 - 3 | 358 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |