Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHSR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204394.1 | NP_989725.1 | 1 - 2 | 1130 | 67 | |
GenBank mRNA | AB095995.1 | - | 1 - 2 | 1130 | 67 | |
AJ309542.1 | - | 1 - 2 | 492 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Recommended Assay Gallus gallus Probe spans exons |