Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204430.2 | NP_989761.2 | 4 - 5 | 1409 | 118 | |
XM_015280410.2 | XP_015135896.1 | 4 - 5 | 1496 | 118 | ||
GenBank mRNA | AJ535692.1 | - | 4 - 5 | 1411 | 117 | |
BX929517.2 | - | 3 - 4 | 1233 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |