Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Gg03365824_m1
See other HNRNPH1 GE Assays ›
Gene Symbol:
HNRNPH1
Gene Name:
heterogeneous nuclear ribonucleoprotein H1
Gene Aliases:
HNRNPH2, HNRPH1
Chromosome Location:
Chr.13: 14281474 - 14298029 on Build GRCg6a
Species:
Chicken
Species Specific ID (Flybase ID):
-
Assay ID
Gg03365824_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: HNRNPH1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_204496.1
NP_989827.1
6 - 7
899
100
XM_015293722.2
XP_015149208.2
5 - 6
840
100
XM_015293723.2
XP_015149209.2
7 - 8
888
100
XM_015293725.2
XP_015149211.2
7 - 8
991
100
XM_015293726.2
XP_015149212.2
6 - 7
828
100
XM_015293728.2
XP_015149214.2
6 - 7
760
100
XM_015293730.2
XP_015149216.2
6 - 7
899
100
XM_025154783.1
XP_025010551.1
6 - 7
899
100
XM_025154784.1
XP_025010552.1
7 - 8
868
100
XM_025154785.1
XP_025010553.1
6 - 7
899
100
XM_025154786.1
XP_025010554.1
6 - 7
688
100
XM_025154787.1
XP_025010555.1
6 - 7
902
100
XM_025154788.1
XP_025010556.1
6 - 7
899
100
XM_025154789.1
XP_025010557.1
6 - 7
929
100
XM_025154790.1
XP_025010558.1
7 - 8
825
100
XM_025154791.1
XP_025010559.1
5 - 6
840
100
XM_025154792.1
XP_025010560.1
6 - 7
807
100
XM_025154793.1
XP_025010561.1
5 - 6
857
100
XM_025154794.1
XP_025010562.1
5 - 6
840
100
XM_025154795.1
XP_025010563.1
6 - 7
899
100
XM_025154796.1
XP_025010564.1
7 - 8
817
100
XM_025154797.1
XP_025010565.1
7 - 8
991
100
GenBank mRNA
AJ719489.1
-
5 - 6
836
100
AJ851662.1
-
6 - 7
800
100
AY278548.1
-
6 - 7
899
100
Back To Top
More Information
Set Membership:
Amplicon length between 86 and 100
Gallus gallus
Probe spans exons
Panther Classification:
Molecular Function -
RNA splicing factor
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0