Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SULT1B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204545.1 | NP_989876.1 | 4 - 5 | 455 | 65 | |
GenBank mRNA | AJ494980.1 | - | 4 - 5 | 474 | 65 | |
BX932004.1 | - | 4 - 5 | 443 | 65 | ||
CR406784.1 | - | 4 - 5 | 437 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |