Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR5A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205077.1 | NP_990408.1 | 3 - 4 | 376 | 98 | |
XM_015279332.2 | XP_015134818.1 | 3 - 4 | 572 | 98 | ||
XM_015279333.2 | XP_015134819.1 | 3 - 4 | 581 | 98 | ||
XM_015279334.2 | XP_015134820.1 | 3 - 4 | 582 | 98 | ||
GenBank mRNA | AB002404.1 | - | 3 - 4 | 376 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |