Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205094.1 | NP_990425.1 | 1 - 2 | 139 | 67 | |
GenBank mRNA | U72992.1 | - | 1 - 2 | 139 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |