Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFAP20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005833.1 | NP_001005833.1 | 3 - 4 | 374 | 94 | |
GenBank mRNA | AJ721091.1 | - | 3 - 4 | 374 | 94 | |
CR386276.1 | - | 3 - 4 | 381 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Gallus gallus Probe spans exons |