Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006153.1 | NP_001006153.1 | 5 - 6 | 510 | 94 | |
XM_025154856.1 | XP_025010624.1 | 5 - 6 | 524 | 94 | ||
XM_025154857.1 | XP_025010625.1 | 5 - 6 | 494 | 94 | ||
GenBank mRNA | AJ720129.1 | - | 5 - 6 | 510 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |