Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006227.1 | NP_001006227.1 | 6 - 7 | 516 | 87 | |
GenBank mRNA | AJ720776.1 | - | 6 - 7 | 548 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Gallus gallus Probe spans exons |