Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT20H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006275.1 | NP_001006275.1 | 18 - 19 | 1727 | 80 | |
XM_015277740.2 | XP_015133226.1 | 19 - 20 | 2126 | 80 | ||
XM_015277745.2 | XP_015133231.1 | 19 - 20 | 2123 | 80 | ||
XM_015277755.2 | XP_015133241.1 | 19 - 20 | 2027 | 80 | ||
XM_015277763.2 | XP_015133249.1 | 19 - 20 | 2123 | 80 | ||
XM_015277765.2 | XP_015133251.1 | 19 - 20 | 2030 | 80 | ||
XM_015277769.2 | XP_015133255.1 | 19 - 20 | 2030 | 80 | ||
XM_015277772.2 | XP_015133258.1 | 19 - 20 | 1787 | 80 | ||
XM_015277779.2 | XP_015133265.1 | 19 - 20 | 1784 | 80 | ||
XM_015277782.2 | XP_015133268.1 | 19 - 20 | 1787 | 80 | ||
XM_015277786.2 | XP_015133272.1 | 19 - 20 | 1876 | 80 | ||
XM_015277793.2 | XP_015133279.1 | 19 - 20 | 2122 | 80 | ||
XM_025150489.1 | XP_025006257.1 | 19 - 20 | 2027 | 80 | ||
XM_025150499.1 | XP_025006267.1 | 19 - 20 | 1787 | 80 | ||
GenBank mRNA | AJ719513.1 | - | 18 - 19 | 1727 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |