Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT20H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006275.1 | NP_001006275.1 | 8 - 9 | 646 | 149 | |
XM_015277740.2 | XP_015133226.1 | 9 - 10 | 799 | 149 | ||
XM_015277745.2 | XP_015133231.1 | 9 - 10 | 796 | 149 | ||
XM_015277755.2 | XP_015133241.1 | 9 - 10 | 703 | 149 | ||
XM_015277763.2 | XP_015133249.1 | 9 - 10 | 796 | 149 | ||
XM_015277765.2 | XP_015133251.1 | 9 - 10 | 703 | 149 | ||
XM_015277769.2 | XP_015133255.1 | 9 - 10 | 703 | 149 | ||
XM_015277772.2 | XP_015133258.1 | 9 - 10 | 703 | 149 | ||
XM_015277779.2 | XP_015133265.1 | 9 - 10 | 703 | 149 | ||
XM_015277782.2 | XP_015133268.1 | 9 - 10 | 703 | 149 | ||
XM_015277786.2 | XP_015133272.1 | 9 - 10 | 795 | 149 | ||
XM_015277793.2 | XP_015133279.1 | 9 - 10 | 795 | 149 | ||
XM_025150489.1 | XP_025006257.1 | 9 - 10 | 703 | 149 | ||
XM_025150499.1 | XP_025006267.1 | 9 - 10 | 703 | 149 | ||
GenBank mRNA | AJ719513.1 | - | 8 - 9 | 646 | 149 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |