Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PECR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006522.1 | NP_001006522.1 | 6 - 7 | 722 | 74 | |
GenBank mRNA | AJ720820.1 | - | 6 - 7 | 722 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |