Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006540.1 | NP_001006540.1 | 2 - 3 | 612 | 66 | |
GenBank mRNA | AJ720228.1 | - | 2 - 3 | 612 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Gallus gallus Probe spans exons |