Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM172A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006574.1 | NP_001006574.1 | 10 - 11 | 1229 | 120 | |
XM_015280521.2 | XP_015136007.1 | 9 - 10 | 1067 | 120 | ||
GenBank mRNA | AJ720240.1 | - | 10 - 11 | 1229 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |