Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006577.2 | NP_001006577.2 | 1 - 2 | 98 | 112 | |
NM_001194996.1 | NP_001181925.1 | 1 - 2 | 98 | 112 | ||
XM_015277311.2 | XP_015132797.1 | 1 - 2 | 115 | 112 | ||
XM_015277312.2 | XP_015132798.1 | 1 - 2 | 129 | 112 | ||
GenBank mRNA | AJ720892.1 | - | 1 - 2 | 99 | 112 | |
BU335726.1 | - | 1 - 2 | 71 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |