Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006577.2 | NP_001006577.2 | 5 - 6 | 482 | 116 | |
NM_001194996.1 | NP_001181925.1 | 5 - 6 | 482 | 116 | ||
XM_015277311.2 | XP_015132797.1 | 5 - 6 | 499 | 116 | ||
XM_015277312.2 | XP_015132798.1 | 5 - 6 | 513 | 116 | ||
GenBank mRNA | AJ720892.1 | - | 5 - 6 | 483 | 116 | |
BU335726.1 | - | 5 - 6 | 455 | 116 | ||
DT656730.1 | - | 3 - 4 | 249 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |