Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM230 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007877.1 | NP_001007878.1 | 2 - 3 | 165 | 80 | |
XM_015297304.2 | XP_015152790.1 | 2 - 3 | 206 | 80 | ||
GenBank mRNA | AJ719760.1 | - | 2 - 3 | 165 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |