Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007892.3 | NP_001007893.1 | 2 - 3 | 609 | 69 | |
GenBank mRNA | BU269517.1 | - | 2 - 3 | 385 | 69 | |
BU327476.1 | - | 2 - 3 | 292 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Gallus gallus Probe spans exons |