Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMTM7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007894.1 | NP_001007895.1 | 3 - 4 | 490 | 138 | |
GenBank mRNA | AJ719741.1 | - | 3 - 4 | 490 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |