Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007917.2 | NP_001007918.1 | 2 - 3 | 394 | 72 | |
GenBank mRNA | AJ720634.1 | - | 2 - 3 | 359 | 72 | |
BU353194.1 | - | 2 - 3 | 398 | 72 | ||
DN929287.1 | - | 2 - 3 | 382 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Gallus gallus Probe spans exons |