Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007975.1 | NP_001007976.1 | 5 - 6 | 910 | 60 | |
GenBank mRNA | AJ719406.1 | - | 5 - 6 | 910 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Gallus gallus Probe spans exons |