Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008444.1 | NP_001008444.1 | 1 - 2 | 429 | 141 | |
XM_015293289.2 | XP_015148775.1 | 2 - 3 | 718 | 141 | ||
XM_015293337.2 | XP_015148823.1 | 2 - 3 | 1041 | 141 | ||
XM_015293371.2 | XP_015148857.1 | 2 - 3 | 680 | 141 | ||
XM_015293418.2 | XP_015148904.1 | 2 - 3 | 718 | 141 | ||
GenBank mRNA | X56931.1 | - | 429 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Gallus gallus Probe spans exons |