Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACTR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008446.1 | NP_001008446.1 | 3 - 4 | 792 | 81 | |
GenBank mRNA | AJ720530.1 | - | 3 - 4 | 792 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Gallus gallus Probe spans exons |