Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008479.1 | NP_001008479.1 | 4 - 5 | 324 | 130 | |
GenBank mRNA | AJ720221.1 | - | 4 - 5 | 324 | 130 | |
BX930917.1 | - | 4 - 5 | 361 | 130 | ||
CR405748.1 | - | 4 - 5 | 372 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |