Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204745.1 | NP_990076.1 | 7 - 8 | 1003 | 111 | |
XM_015291975.1 | XP_015147461.1 | 7 - 8 | 1008 | 111 | ||
GenBank mRNA | AF237950.2 | - | 7 - 8 | 1003 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |