Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205113.1 | NP_990444.1 | 2 | 354 | 87 | |
GenBank mRNA | D31864.1 | - | 2 | 354 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |