Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143931.1 | NP_001137403.1 | 14 - 15 | 1566 | 103 | |
XM_015283362.2 | XP_015138848.1 | 13 - 14 | 1397 | 103 | ||
GenBank mRNA | EU344154.1 | - | 14 - 15 | 1566 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Gallus gallus Probe spans exons |