Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204520.2 | NP_989851.2 | 3 | 1046 | 111 | |
XM_015287272.2 | XP_015142758.1 | 5 | 1389 | 111 | ||
GenBank mRNA | AY600957.1 | - | 3 | 1046 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Gallus gallus Both primers and probe map within a single exon |