Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249275.1 | NP_001236204.1 | 3 - 4 | 314 | 90 | |
GenBank mRNA | AJ239132.1 | - | 3 - 4 | 314 | 90 | |
BT097689.1 | - | 3 - 4 | 332 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |