Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWINV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249396.1 | NP_001236325.1 | 4 - 5 | 1638 | 96 | |
GenBank mRNA | AJ563368.1 | - | 4 - 5 | 1638 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Glycine max Probe spans exons |