Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249748.1 | NP_001236677.1 | 4 - 5 | 596 | 83 | |
GenBank mRNA | AK244377.1 | - | 4 - 5 | 627 | 83 | |
AK286765.1 | - | 4 - 5 | 653 | 83 | ||
AY151271.1 | - | 4 - 5 | 596 | 83 | ||
AY151272.1 | - | 4 - 5 | 596 | 83 | ||
AY151273.1 | - | 4 - 5 | 596 | 83 | ||
AY151274.1 | - | 4 - 5 | 596 | 83 | ||
AY151275.1 | - | 4 - 5 | 596 | 83 | ||
GQ422778.1 | - | 4 - 5 | 596 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Glycine max Probe spans exons |