Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P5CS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251224.1 | NP_001238153.1 | 18 - 19 | 1994 | 101 | |
GenBank mRNA | AY492005.1 | - | 18 - 19 | 1994 | 101 | |
FM999729.1 | - | 18 - 19 | 1994 | 101 | ||
FM999730.1 | - | 18 - 19 | 1994 | 101 | ||
FN564570.1 | - | 18 - 19 | 1994 | 101 | ||
FN564571.1 | - | 18 - 19 | 1994 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Glycine max Probe spans exons |