Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248792.1 | NP_001235721.1 | 2 - 3 | 369 | 77 | |
GenBank mRNA | AK287395.1 | - | 2 - 3 | 381 | 77 | |
U77678.1 | - | 2 - 3 | 369 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |