Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249944.1 | NP_001236873.1 | 6 - 7 | 621 | 69 | |
GenBank mRNA | X95582.1 | - | 6 - 7 | 621 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Glycine max Probe spans exons |