Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AM158274.1 | - | 2 - 3 | 357 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Glycine max Probe spans exons |