Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251288.1 | NP_001238217.1 | 9 - 10 | 1020 | 101 | |
GenBank mRNA | AY856369.1 | - | 9 - 10 | 1020 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Glycine max Probe spans exons |