Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249926.1 | NP_001236855.1 | 3 | 1050 | 103 | |
GenBank mRNA | DQ028769.1 | - | 3 | 1050 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Glycine max Both primers and probe map within a single exon |