Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYB98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001248936.1 | NP_001235865.1 | 3 | 552 | 104 | |
GenBank mRNA | DQ822907.1 | - | 3 | 552 | 104 |
Gene Symbol: LOC100306400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251334.1 | NP_001238263.1 | 3 | 567 | 104 | |
GenBank mRNA | BT090480.1 | - | 3 | 567 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Glycine max Both primers and probe map within a single exon |