Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041501.hg.1 |
TC02005050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041501.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041501.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041506.hg.1 |
TC02005050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041506.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041506.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041505.hg.1 |
TC02005050.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041505.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041505.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02041505.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228597.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228597.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228597.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228597.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228597.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200016755.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228598.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228598.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228598.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228598.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228598.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228596.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228596.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228596.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228596.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228596.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228601.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228602.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228602.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228602.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228602.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228602.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
ENST00000295108.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
ENST00000496876.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
NM_002500.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200228600.hg.1 |
TC0200016755.hg.1 |
Clariom D TC |