Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_002762.3 | NP_002753.2 | 1 - 2 | 387 | 89 | |
NR_104428.1 | 1 - 2 | 387 | 89 | |||
GenBank mRNA | AK130957.1 | - | 1 - 2 | 387 | 89 | |
AK292447.1 | - | 1 - 2 | 387 | 89 | ||
AK311798.1 | - | 1 - 2 | 387 | 89 | ||
BC005303.1 | - | 1 - 2 | 387 | 3935273 | 89 | |
BC042671.1 | - | 1 - 2 | 356 | 5166075 | 89 | |
BC066338.1 | - | 1 - 2 | 387 | 4821351 | 89 | |
BP368978.1 | - | 1 - 2 | 387 | 89 | ||
X07862.1 | - | 1 - 2 | 358 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927252 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16851396 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439227 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439220 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439241 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927247 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927256 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927249 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927251 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439233 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16851394 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439230 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927258 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30439240 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927246 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927253 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09927242 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3358893 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_24_P82240 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2281 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_2282 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 43363 | - | - |
Agilent Human Genome CGH Microarray 44B | A_14_P115650 | - | - |
Agilent Human Genome CGH Microarray 44A | A_14_P115650 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48033 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 60365 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3358893 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_19_P00813586 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3358893 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439240 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439227 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927253 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439241 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927242 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439220 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851393 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439234 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927253 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927258 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439230 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851394 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439241 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927242 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16851396 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09927246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439240 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30439227 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_24_P82240 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_14_P115650 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927246 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927249 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927259 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927256 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927255 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927244 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927258 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927252 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P09927253 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09927247 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16851388 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P16851389 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09927246 | - | - |
Agilent Human CpG Island ChIP-on-Chip Microarray 244K (G4492A) | A_17_P09927248 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_14_P115650 | - | - |
Agilent Human Genome CGH Microarray 4x44K | A_14_P115650 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P82240 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 173329 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 101267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927261 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927272 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927201 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927205 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927211 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16851396 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927270 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927207 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927268 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927214 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927232 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927265 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927244 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927216 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927256 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927190 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927249 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927280 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927251 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927276 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927267 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927246 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16851390 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16851381 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927259 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927195 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927275 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927278 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927273 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927196 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927255 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927226 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927253 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927263 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927209 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927203 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927228 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927258 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927197 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927194 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927281 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09927223 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_002762_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7999439 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7999438 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7999437 | - | - |
Affymetrix Human Gene 1.0 ST Array | 7999436 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7999435 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 210122_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11755740_a_at | - | - |
Affymetrix Human Genome U219 Array | 11755740_PM_a_at | - | - |
Human Genome U133 Plus 2.0 Array | 210122_at | - | - |
Human Genome U133 Plus 2.0 Array | 210122_PM_at | - | - |
Human Genome U133A 2.0 Array | 210122_PM_at | - | - |
Human Genome U133A 2.0 Array | 210122_at | - | - |
Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NM_002762_at | - | - |
AffymetrixPathwork Diagnostics Pathchip | pw509655 | - | - |
Human Genome U95 Set | 34658_PM_at | - | - |
Human Genome U95 Set | 34658_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 210122_at9 | - | - |
Human Genome U133 Set | 210122_PM_at | - | - |
Human Genome U133 Set | 210122_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053484 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053484 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1681772 | CR609803.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1681772 | CR621790.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1681772 | X07862.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1681772 | CR603750.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4506110-S | CR609803.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4506110-S | CR621790.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4506110-S | X07862.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4506110-S | CR603750.1 | human_transcriptome |
Sentrix Human-6 Expression BeadChip | GI_4506110-S | BC066338.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_2715-S | - | - |
Illumina human-6 v1.0 expression beadchip | 0004280538 | - | - |
Illumina humanRef-8 v1.0 expression beadchip | 0004280538 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 5360164 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_2715 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1681772 | BC066338.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1681772 | CR609803.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1681772 | CR621790.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1681772 | X07862.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1681772 | CR603750.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1681772 | BC066338.1 | human_transcriptome |