Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
TC07000137.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000258743.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000401630.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000401651.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000404625.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000407492.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
ENST00000420258.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07002126.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700006890.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000258743.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000401630.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000401651.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000404625.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000406575.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000407492.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000426291.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000464710.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
ENST00000485300.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
NM_000600.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700147663.hg.1 |
TC0700006890.hg.1 |
Clariom D TC |