Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
TC03001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
ENST00000264246.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
ENST00000383668.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
ENST00000478182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030377.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
TC03001684.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
ENST00000264246.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
ENST00000383668.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
ENST00000478182.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03030378.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190206.hg.1 |
TC0300012123.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300190207.hg.1 |
TC0300012123.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000264246.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000383669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000463729.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
ENST00000478182.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300012123.hg.1 |
NM_005191.3 |
Refseq |