Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000320027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000380405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000445273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000602786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000626866.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000630972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_006644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000320027.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000380405.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000445273.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000602786.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000626866.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
ENST00000630972.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286503.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286504.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_001286505.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
NM_006644.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157557.hg.1 |
TC1300008533.hg.1 |
Clariom D TC |