Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
TC11003440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
ENST00000528474.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023765.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023768.hg.1 |
TC11003440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023768.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023768.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023769.hg.1 |
TC11003440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023769.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023769.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023770.hg.1 |
TC11003440.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023770.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11023770.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204749.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204749.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204749.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204749.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204749.hg.1 |
TC1100013192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100013192.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
ENST00000533438.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
NM_001166212.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204744.hg.1 |
TC1100013192.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100204748.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204748.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204748.hg.1 |
ENST00000312438.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100204748.hg.1 |
NM_013246.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100204748.hg.1 |
TC1100013192.hg.1 |
Clariom D TC |