Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001038.hg.1 |
TC6_dbb_hap3000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001038.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001038.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_dbb_hap3001038.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001076.hg.1 |
TC6_qbl_hap6000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001076.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001076.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001076.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000910.hg.1 |
TC6_mcf_hap5000058.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000910.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000910.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5000910.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000886.hg.1 |
TC6_ssto_hap7000059.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000886.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000886.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7000886.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
TC06000377.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
ENST00000375918.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
ENST00000375920.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06003736.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001095.hg.1 |
TC6_cox_hap2000072.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001095.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001095.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001095.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001029.hg.1 |
TC6_mann_hap4000063.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001029.hg.1 |
NM_001256169.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001029.hg.1 |
NM_019101.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001029.hg.1 |
NR_045828.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151905.hg.1 |
TC0600007602.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600007602.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375916.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375918.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
ENST00000375920.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600151907.hg.1 |
TC0600007602.hg.1 |
Clariom D TC |