Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001603.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001603.hg.1 |
ENST00000380390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001603.hg.1 |
ENST00000477339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001603.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001603.hg.1 |
TC11000099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001604.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001604.hg.1 |
ENST00000380390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001604.hg.1 |
ENST00000477339.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001604.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001604.hg.1 |
TC11000099.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001602.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001602.hg.1 |
ENST00000380390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001602.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11001602.hg.1 |
TC11000099.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146679.hg.1 |
TC1100006666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146680.hg.1 |
TC1100006666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100146678.hg.1 |
TC1100006666.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000300762.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000380390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
ENST00000477339.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006666.hg.1 |
NM_021801.4 |
Refseq |